Sodium chloride, 5M aq. soln., RNAse free
Sodium chloride, 5M aq. soln., RNAse free
Sodium chloride, 5M aq. soln., RNAse free
Thermo Scientific Chemicals

Sodium chloride, 5M aq. soln., RNAse free

CAS: 7647-14-5 | ClNa | 58.44 g/mol
製品番号(カタログ番号)数量
J60434.AK
または、製品番号J60434-AK
250 mL
製品番号(カタログ番号) J60434.AK
または、製品番号J60434-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
さらに表示
CompositionSodium Chloride: 5M
CommentRNase free
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

Solubility
Soluble in water (358 mg/ml at 20°C), alcohol (very slightly), glycerol (100 mg/ml), and ammonia. Insoluble in hydrochloric acid.

Notes
Incompatible with strong oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances. Store protected from moisture.
RUO – Research Use Only

General References:

  1. Guangling Song.; Andrej Atrens.; Xianliang Wu.; Bo Zhang. Corrosion behaviour of AZ21, AZ501 and AZ91 in sodium chloride. Corros. Sci.. 1998, 40 (10), 1769-1791.
  2. D. H. Jennings. The Effects Of Sodium Chloride On Higher Plants. Biological Reviews. 1976, 51 (4), 453-486.