Potassium phosphate, 0.5M buffer soln., pH 7.0
Potassium phosphate, 0.5M buffer soln., pH 7.0
Potassium phosphate, 0.5M buffer soln., pH 7.0
Thermo Scientific Chemicals

Potassium phosphate, 0.5M buffer soln., pH 7.0

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J60584.AK
または、製品番号J60584-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS7778-77-0
化学物質名または材質Potassium phosphate
IUPAC名potassium dihydrogen phosphate
InChIキーGNSKLFRGEWLPPA-UHFFFAOYSA-M
MDL番号MFCD00011401 MFCD00147253
さらに表示
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate buffer is typically used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.

Solubility
Soluble in water, Insoluble to slightly soluble in ethanol, insoluble in alcohol.

Notes
Store sealed bottle containing the dehydrated medium at 2 - 30C. Once opened and recapped, place container in a low humidity environment at the same storage temperature. Protect from moisture and light by keeping container tightly closed.
RUO – Research Use Only

General References:

  1. Pardue, K., and Williams, D. Quantitative determination of non-ionic surfactants in protein samples, using ion-exchange guard columns. Biotechniques. 1993, 14(4), 580-583.
  2. Pikal-Cleland, K.A., et al. Protein denaturation during freezing and thawing in phosphate buffer systems: monobasic and tetrameric beta-galactosidase. . Arch. Biochem. Biophys. 2000, 384(2), 398-406.