Thermo Scientific Chemicals

Ginkgolide B, Thermo Scientific Chemicals

Catalog number: J60646.MA
10 mg, Each
Thermo Scientific Chemicals

Ginkgolide B, Thermo Scientific Chemicals

Catalog number: J60646.MA
10 mg, Each
Quantity
Catalog number: J60646.MA
also known as J60646-MA
Price (USD)
Price: 112.00
Online price: 97.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15291-77-7
Assay (unspecified)
98.0% min.

Description

Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

Solubility
Soluble in DMSO. Slightly soluble in water and ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text