Citrate, 0.5M buffer soln., pH 5.5
Citrate, 0.5M buffer soln., pH 5.5
Citrate, 0.5M buffer soln., pH 5.5
Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 5.5

CAS: 6132-04-3294.10 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J60754.AK
または、製品番号J60754-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6132-04-3
IUPAC Nametrisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular FormulaC6H9Na3O9
InChI KeyNLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILESO.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
さらに表示
Appearance (Color)Clear colorless
FormLiquid
pH5.35-5.65
Concentration500 mM
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only