Thermo Scientific Chemicals

Nitrendipine, Thermo Scientific Chemicals

Catalog number: J60841.ME
500 mg, Each
Thermo Scientific Chemicals

Nitrendipine, Thermo Scientific Chemicals

Catalog number: J60841.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60841.ME
also known as J60841-ME
Price (USD)
Price: 369.00
Online price: 318.65
Your price:
Quantity
-

Chemical Identifiers

CAS
39562-70-4
IUPAC Name
3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C18H20N2O6
InChI Key
PVHUJELLJLJGLN-UHFFFAOYNA-N
SMILES
CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC
Form
Powder
Assay from Supplier's CofA
≥95.0%

Description

Nitrendipine modulates NMDA receptor channel function in mammalian neurons. Nitrendipine has been shown to inhibit neutrophil adhesion to vascular endothelium. It is used in the treatment of primary hypertension to decrease blood pressure. Nitrendipine is used alone or with an angiotensin-converting enzyme inhibitor, to treat hypertension, chronic stable angina pectoris, and Prinzmetal's variant angina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO (17.5 mg/ml), methanol (15 mg/ml), ethyl acetate, and ethanol (25 mM). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text