Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.5%
Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.5%
Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.5%
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, Electrophoresis Grade, 99.5%

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
製品番号(カタログ番号) J61016.22
または、製品番号J61016-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
さらに表示
Protease activityNone detected
RNaseNone detected
Trace MetalsFe: 5ppm max.
Trace MetalsPb: 5ppm max.
Insoluble material0.005% max.
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only