Thioflavine T, tech. 75%
Thioflavine T, tech. 75%
Thioflavine T, tech. 75%
Thermo Scientific Chemicals

Thioflavine T, tech. 75%

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease | CAS: 2390-54-7 | C17H19ClN2S | 318.86 g/mol
製品番号(カタログ番号) J61043.14
または、製品番号J61043-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2390-54-7
IUPAC Name2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
Molecular FormulaC17H19ClN2S
InChI KeyJADVWWSKYZXRGX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1
さらに表示
FormPowder
Assay (special)Lambda max in water: 409-415 nm
Solution TestSolubility: Conforms
Appearance (Color)Yellow to orange to brown
Identification (FTIR)Conforms
A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only