(+/-)-Atenolol
(+/-)-Atenolol
(+/-)-Atenolol
Thermo Scientific Chemicals

(+/-)-Atenolol

CAS: 29122-68-7 | C14H22N2O3 | 266.341 g/mol
製品番号(カタログ番号)数量
J61199.03
または、製品番号J61199-03
1 g
製品番号(カタログ番号) J61199.03
または、製品番号J61199-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS29122-68-7
IUPAC Name2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
Molecular FormulaC14H22N2O3
InChI KeyMETKIMKYRPQLGS-UHFFFAOYNA-N
SMILESCC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1
さらに表示
Appearance (Color)White to off-white
FormPowder
Assay (unspecified)98.0% min.
Cardioselective beta-adrenergic blocker. An antihypertensive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cardioselective beta-adrenergic blocker
  • An antihypertensive
RUO – Research Use Only