Adrenocorticotropic Hormone (1-13)
Adrenocorticotropic Hormone (1-13)
Adrenocorticotropic Hormone (1-13)
Thermo Scientific Chemicals

Adrenocorticotropic Hormone (1-13)

数量:
0.5 mg
1 mg
5 mg
製品番号(カタログ番号) J61247.MCR
または、製品番号J61247-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
仕様
CAS22006-64-0
フォーマットPowder
IUPAC名(4S)-4-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-[({[(2S)-6-amino-1-[(2S)-2-{[(1S)-1-carboxy-2-methylpropyl]carbamoyl}pyrrolidin-1-yl]-1-oxohexan-2-yl]carbamoyl}methyl)carbamoyl]-2-(1H-indol-3-yl)ethyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-2-phenylethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-amino-3-hydroxypropanamido]-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido]-4-(methylsulfanyl)butanamido]butanoic acid
InChIキーVGBVAARMQYYITG-DESRROFGSA-N
分子式C75H106N20O19S
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
Store at -20°C