4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals

Substrate for alkaline and acid phosphatase determinations
製品番号(カタログ番号)数量
J61401.KN100 Pc.
製品番号(カタログ番号) J61401.KN
価格(JPY)
-
数量:
100 Pc.
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン5324661
CAS4264-83-9
化学物質名または材質4-Nitrophenyl phosphate disodium salt hexahydrate
Yellow
EINECS番号224-246-5
さらに表示
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

Solubility
It is soluble in water.

Notes
The tablets should be stored -20°C
RUO – Research Use Only

General References:

  1. GN Bowers, RB McComb, A Upretti. 4-nitrophenyl phosphate--characterization of high-purity materials for measuring alkaline phosphatase activity in human serum. Clinical chemistry. 1981, 27, (1), 135-143.
  2. R Rej.; JP Bretaudiere. Effects of metal ions on the measurement of alkaline phosphatase activity. Clinical chemistry. 1980, 26, (3), 423-428.