Thermo Scientific Chemicals

Emodin, Thermo Scientific Chemicals

Catalog number: J61600.MD
250 mg, Each
Thermo Scientific Chemicals

Emodin, Thermo Scientific Chemicals

Catalog number: J61600.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61600.MD
also known as J61600-MD
Price (USD)
Quantity
-

Chemical Identifiers

CAS
518-82-1
IUPAC Name
1,3,8-trihydroxy-6-methyl-9,10-dihydroanthracene-9,10-dione
Molecular Formula
C15H10O5
InChI Key
RHMXXJGYXNZAPX-UHFFFAOYSA-N
SMILES
CC1=CC(O)=C2C(=O)C3=C(O)C=C(O)C=C3C(=O)C2=C1
Loss on Drying
1.0% max.
Appearance (Color)
Pale brown
Form
Powder
Assay (HPLC)
90.0% min.
Ash
1.0% max.

Description

A protein tyrosine kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text