Sodium citrate, 0.5M buffer soln., pH 6.0
Thermo Scientific Chemicals

Sodium citrate, 0.5M buffer soln., pH 6.0

数量:
250 mL
500 mL
製品番号(カタログ番号) J61815.AK
または、製品番号J61815-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
化学物質名または材質Sodium citrate buffer soln.
Colorless
健康被害1Warning
健康被害2GHS H Statement
H319
Causes serious eye irritation.
健康被害3GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
さらに表示
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.

Solubility
Fully miscible in water.

Notes
Incompatible with heat and oxidizing agents.
RUO – Research Use Only

General References:

  1. Mostafa Mesbah; Usha Premachandran; William B. Whitman. Precise Measurement of the G+C Content of Deoxyribonucleic Acid by High-Performance Liquid Chromatography. Int J Syst Evol Microbiol.1989, 39, 159-167.
  2. Tomas Lindahl; Annika Andersson. Rate of chain breakage at apurinic sites in double-stranded deoxyribonucleic acid. Biochemistry.1972, 11 , (19), 3618-3623.