L-Methionine, Cell Culture Reagent
L-Methionine, Cell Culture Reagent
L-Methionine, Cell Culture Reagent
L-Methionine, Cell Culture Reagent
Thermo Scientific Chemicals

L-Methionine, Cell Culture Reagent

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol
数量:
50 g
100 g
500 g
製品番号(カタログ番号) J61904.18
または、製品番号J61904-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS63-68-3
IUPAC Name(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular FormulaC5H11NO2S
InChI KeyFFEARJCKVFRZRR-BYPYZUCNSA-N
SMILESCSCC[C@H](N)C(O)=O
さらに表示
Assay from Supplier's CofA≥99.0 to ≤ 101.0%
CommentEndotoxin: ≤6.0EU/g
Impurity contentIron, Fe: ≤10ppm
Optical Rotation@ 20?C: +23.0 to +24.0? (c=2, 6N HCl, dry weight basis)
Heavy Metals (as Pb)≤10ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only