Thermo Scientific Chemicals

Metoprolol tartrate, 98+%, Thermo Scientific Chemicals

Catalog number: J61920.14
25 g, Each
Thermo Scientific Chemicals

Metoprolol tartrate, 98+%, Thermo Scientific Chemicals

Catalog number: J61920.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61920.14
also known as J61920-14
Price (USD)
Price: 322.00
Online price: 307.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56392-17-7
IUPAC Name
(2R,3R)-2,3-dihydroxybutanedioic acid; bis(1-[4-(2-methoxyethyl)phenoxy]-3-[(propan-2-yl)amino]propan-2-ol)
Molecular Formula
C34H56N2O12
InChI Key
YGULWPYYGQCFMP-CEAXSRTFSA-N
SMILES
O[C@H]([C@@H](O)C(O)=O)C(O)=O.COCCC1=CC=C(OCC(O)CNC(C)C)C=C1.COCCC1=CC=C(OCC(O)CNC(C)C)C=C1
Loss on Drying
≤0.5%
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥98.0%
Form
Powder or crystals or crystalline powder
Proton NMR
Conforms to structure

Description

Metoprolol tartrate is an antagonist of the β1-AR (β1-adrenoceptor). It inhibits spontaneous endothelin-1 production in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metoprolol tartrate is an antagonist of the β1-AR (β1-adrenoceptor). It inhibits spontaneous endothelin-1 production in vitro and displays antihypertensive.

Solubility
Soluble in water (>1000 mg/ml), methanol (>500 mg/ml), chloroform (496 mg/ml), ethanol (31 mg/ml at 25°C), and DMSO (100 mg/ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text