Thermo Scientific Chemicals

L-Serine, Cell Culture Reagent

Catalog number: J62187.36
500 g, Each
Thermo Scientific Chemicals

L-Serine, Cell Culture Reagent

Catalog number: J62187.36
500 g, Each
Quantity
Catalog number: J62187.36
also known as J62187-36
Price (USD)
Quantity
-

Description

It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

Solubility
Soluble in water.

Notes
Store in cool, dry place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text