SOB medium, contains 20g/L tryptone, 5g/L yeast extract, 0.95g/L MgCl{2}, 0.5g/L NaCl, and 0.186g/L KCl
Thermo Scientific Chemicals
SOB medium, contains 20g/L tryptone, 5g/L yeast extract, 0.95g/L MgCl{2}, 0.5g/L NaCl, and 0.186g/L KCl
SOB Medium was developed by Hanahan as a nutritionally rich growth medium for preparation and transformation of competent cells. Growth of E. coli in SOB medium results in higher transformation efficiencies of plasmids.
数量:
100 mL
250 mL
製品番号(カタログ番号) J62210.AE
または、製品番号J62210-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
仕様
パッケージングタイプPlastic Bottle
数量100 mL
推奨保存方法Ambient temperatures
形状Liquid
Unit SizeEach
SOB Medium was developed by Hanahan as a nutritionally rich growth medium for preparation and transformation of competent cells. Growth of E. coli in SOB medium results in higher transformation efficiencies of plasmids.