Cytidine-5'-triphosphate disodium salt, 97%
Cytidine-5'-triphosphate disodium salt, 97%
Cytidine-5'-triphosphate disodium salt, 97%
Thermo Scientific Chemicals

Cytidine-5'-triphosphate disodium salt, 97%

CAS: 36051-68-0 | C9H14N3Na2O14P3 | 527.12 g/mol
数量:
500 mg
1 g
2 g
製品番号(カタログ番号) J62238.ME
または、製品番号J62238-ME
価格(JPY)
-
数量:
500 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS36051-68-0
IUPAC Namedisodium phosphono ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC9H14N3Na2O14P3
InChI KeyNFQMDTRPCFJJND-WFIJOQBCSA-L
SMILES[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
さらに表示
Heavy Metals≤10ppm (non-U.S. specification)
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (unspecified)≥88.0% (dry wt. basis)
Assay (HPLC)≥96.0%
さらに表示
Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhou, H.; Kong, S.; Pan, Y.; Zhang, Z.; Deng, L. Microwave-assisted fabrication of strontium doped apatite coating on Ti6Al4V. Mater. Sci. Eng., C 2015, 56, 174-180.
  2. Zheng, S.; Lu, J.; Li, W.; Qin, Y.; Yan, D.; Evans, D. G.; Duan, X. The 2-phenylbenzimidazole-5-sulfonate/layered double hydroxide co-intercalation composite and its luminescence response to nucleotides. J. Mater. Chem. C 2014, 2 (26), 5161-5167.