Thermo Scientific Chemicals

Maprotiline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J62321.06
5 g, Each
Thermo Scientific Chemicals

Maprotiline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: J62321.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62321.06
also known as J62321-06
Price (USD)
Price: 375.00
Online price: 327.65
Your price:
Quantity
-

Description

Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.

Solubility
Soluble to 50mg/ml in water. Soluble in ethanol at 10mM

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text