beta-Endorphin, human
Thermo Scientific Chemicals

beta-Endorphin, human

3465.06 g/mol
製品番号(カタログ番号)数量
J62374.MCR
または、製品番号J62374-M^
1 mg
製品番号(カタログ番号) J62374.MCR
または、製品番号J62374-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS61214-51-5
Assay (HPLC)≥95.0%
FormLyophilized powder
Molecular WeightTheoretical MW: 3465.10
beta-endorphins produce analgesia by binding to opioid receptors (particularly of the mu subtype) at both pre- and post- synaptic nerve terminals, primarily exerting their effect through presynaptic binding. In the central nervous system, beta-endorphins similarly bind mu-opioid receptors and exert their primary action at presynaptic nerve terminals. It is a potent pain suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only