TE buffer, (20X), pH 8.0, autoclaved
TE buffer, (20X), pH 8.0, autoclaved
TE buffer, (20X), pH 8.0, autoclaved
Thermo Scientific Chemicals

TE buffer, (20X), pH 8.0, autoclaved

Commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids. | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
数量:
250 mL
500 mL
製品番号(カタログ番号) J62388.AK
または、製品番号J62388-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS1185-53-1
化学物質名または材質TE buffer
IUPAC名2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChIキーQKNYBSVHEMOAJP-UHFFFAOYSA-N
MDL番号MFCD00236359
さらに表示
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only