Polyvinylpyrrolidone, M.W. 40,000
Polyvinylpyrrolidone, M.W. 40,000
Polyvinylpyrrolidone, M.W. 40,000
Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 40,000

A cryoprotectant, also useful in plant cell culture research | CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
製品番号(カタログ番号) J62417.A1
または、製品番号J62417-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
Appearance (Color)White to yellow
pH3.0-7.0
Assay (special)K value: 27-32
Residue on ignition≤0.1%
Heavy Metals (as Pb)≤10 ppm
さらに表示
A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only