Penicillin V potassium salt
Penicillin V potassium salt
Penicillin V potassium salt
Thermo Scientific Chemicals

Penicillin V potassium salt

Inhibits bacterial cell wall synthesis | CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol
数量:
10 g
25 g
100 g
製品番号(カタログ番号) J62442.22
または、製品番号J62442-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS132-98-9
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥98.0%
Mass SpecConsistent with structure
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

Solubility
Soluble in water, dimethyl sulfoxide, and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
RUO – Research Use Only

General References:

  1. Li-Li Du, Qi Wu.; Chun-Xiu Chen.; Bo-Kai Liu.; Xian-Fu Lin. A two-step, one-pot enzymatic synthesis of ampicillin from penicillin G potassium salt.J. Mol. Catal. B: Enzym. 2009, 58 (1-4),208-211 .
  2. John C. Sheehan.; Kenneth R. Henery-Logan. The Total Synthesis of Penicillin V.J. Am. Chem. Soc. 1959, 81 (12),3089-3094 .