TRIS-buffered saline (TBS, 10X, high salt) pH 7.4
TRIS-buffered saline (TBS, 10X, high salt) pH 7.4
TRIS-buffered saline (TBS, 10X, high salt) pH 7.4
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X, high salt) pH 7.4

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
数量:
500 mL
1 L
2 L
製品番号(カタログ番号) J62664.AP
または、製品番号J62664-AP
価格(JPY)
-
数量:
500 mL
一括またはカスタム形式をリクエストする
仕様
CAS77-86-1
ChEBICHEBI:9754
化学物質名または材質TRIS-buffered saline
IUPAC名2-amino-2-(hydroxymethyl)propane-1,3-diol
InChIキーLENZDBCJOHFCAS-UHFFFAOYSA-N
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only