Thermo Scientific Chemicals

Dibucaine hydrochloride, Thermo Scientific Chemicals

Catalog number: J62804.06
5 g, Each
Thermo Scientific Chemicals

Dibucaine hydrochloride, Thermo Scientific Chemicals

Catalog number: J62804.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62804.06
also known as J62804-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
61-12-1
IUPAC Name
{2-[(2-butoxyquinolin-4-yl)formamido]ethyl}diethylazanium chloride
Molecular Formula
C20H30ClN3O2
InChI Key
IVHBBMHQKZBJEU-UHFFFAOYSA-N
SMILES
[Cl-].CCCCOC1=CC(C(=O)NCC[NH+](CC)CC)=C2C=CC=CC2=N1
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥97.0%
Form
Powder or crystals or crystalline powder

Description

It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

Solubility
Freely soluble in water, alcohol, acetone, or chloroform

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text