Thermo Scientific Chemicals

L-Asparagine monohydrate, Cell Culture Reagent

Catalog number: J62869.A1
1 kg, Each
Thermo Scientific Chemicals

L-Asparagine monohydrate, Cell Culture Reagent

Catalog number: J62869.A1
1 kg, Each
Quantity
Catalog number: J62869.A1
also known as J62869-A1
Price (USD)
Price: 583.00
Online price: 504.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5794-13-8
IUPAC Name
(2S)-2-amino-3-carbamoylpropanoic acid hydrate
Molecular Formula
C4H10N2O4
InChI Key
RBMGJIZCEWRQES-DKWTVANSSA-N
SMILES
O.N[C@@H](CC(N)=O)C(O)=O
Appearance (Color)
White
Form
Crystals or crystalline powder
Assay (unspecified)
98.5-101.0%
Heavy Metals (as Pb)
≤10 ppm
pH
4.4-6.4

Description

It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text