Thermo Scientific Chemicals

myo-Inositol, Cell Culture Grade

Catalog number: J62886.22
100 g, Each
Thermo Scientific Chemicals

myo-Inositol, Cell Culture Grade

Catalog number: J62886.22
100 g, Each
Quantity
Catalog number: J62886.22
also known as J62886-22
Price (USD)
Price: 144.00
Online price: 123.65
Your price:
Quantity
-

Chemical Identifiers

CAS
87-89-8
IUPAC Name
cyclohexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H12O6
InChI Key
CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES
OC1C(O)C(O)C(O)C(O)C1O
Form
Solid
Loss on Drying
≤ 0.5 %
Appearance (Color)
White
Assay (HPLC)
≥ 97.0 %
Heavy Metals
≤ 10 ppm

Description

It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

Solubility
It is soluble in water (50 mg/ml).

Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text