Neohesperidin dihydrochalcone hydrate, 98+%
Neohesperidin dihydrochalcone hydrate, 98+%
Neohesperidin dihydrochalcone hydrate, 98+%
Thermo Scientific Chemicals

Neohesperidin dihydrochalcone hydrate, 98+%

Artificial sweetener derived from citrus | CAS: 20702-77-6 | C28H36O15 | 612.58 g/mol
製品番号(カタログ番号) J62922.03
または、製品番号J62922-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20702-77-6
IUPAC Name1-(4-{[4,5-dihydroxy-6-(hydroxymethyl)-3-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}-2,6-dihydroxyphenyl)-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
Molecular FormulaC28H36O15
InChI KeyITVGXXMINPYUHD-UHFFFAOYSA-N
SMILESCOC1=C(O)C=C(CCC(=O)C2=C(O)C=C(OC3OC(CO)C(O)C(O)C3OC3OC(C)C(O)C(O)C3O)C=C2O)C=C1
さらに表示
Assay (Titration)98.0% min.
Loss on Drying11.0% max.
Neohesperidin dihydrochalcone hydrate, is a flavonoid sweetening agent with potent antioxidant activity. It is antioxidant agent. artificial sweetener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neohesperidin dihydrochalcone hydrate, is a flavonoid sweetening agent with potent antioxidant activity. It is antioxidant agent. artificial sweetener.

Solubility
Soluble in water to 100 mM (with heating).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. L Krbechek.; GE Inglett.; M Holik.; B Dowling. Dihydrochalcones. Synthesis of potential sweetening agents. J. Agric. Food Chem. 196816 (1), 108-112.
  2. W Shin.; SJ Kim.; JM Shin.; SH Kim. Structure-taste correlations in sweet dihydrochalcone, sweet dihydroisocoumarin, and bitter flavone compounds. Journal of medicinal chemistry. 199538 (21), 4325-4331.