Thermo Scientific Chemicals

Folic acid, 95-102%, Thermo Scientific Chemicals

Catalog number: J62937.14
25 g, Each
Thermo Scientific Chemicals

Folic acid, 95-102%, Thermo Scientific Chemicals

Catalog number: J62937.14
25 g, Each
Quantity
Catalog number: J62937.14
also known as J62937-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
59-30-3
IUPAC Name
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Molecular Formula
C19H19N7O6
InChI Key
OVBPIULPVIDEAO-LBPRGKRZSA-N
SMILES
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
Appearance (Color)
Yellow to orange
Form
Crystalline powder
Assay (unspecified)
95.0 to 102.0%
Assay (UV/vis)
A256/A365: 2.8 to 3.0
Residue on ignition
≤ 0.3%

Description

Folic acid and its derivatives are essential mediators of one-carbon metabolism within cells.It is especially important in aiding rapid cell division and growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text