Neutral Red
Neutral Red
Neutral Red
Thermo Scientific Chemicals

Neutral Red

Nuclear counterstain useful in pH range 6.8 to 8.0 | CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol
製品番号(カタログ番号) J63054.03
または、製品番号J63054-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS553-24-2
IUPAC Namehydrogen N2,N2,7-trimethylphenazine-2,8-diamine chloride
Molecular FormulaC15H17ClN4
InChI KeyPGSADBUBUOPOJS-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=NC2=C1
さらに表示
Solution TestSolubility 0.1% (50% ethanol and 2 ml acetic acid): Clear red solution
Appearance (Color)Very dark green
Assay (special)Dye content: ≥ 50%
FormPowder
Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only