L-(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-(-)-Malic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number J63221.A4
also known as J63221-A4
Price (USD)/ Each
168.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
198.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)/ Each
168.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
198.00 
Save 29.35 (15%)
Add to cart
L-(-)-Malic acid, 99%, Thermo Scientific Chemicals
Catalog numberJ63221.A4
Price (USD)/ Each
168.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
198.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
CAS97-67-6
IUPAC Name(2S)-2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-REOHCLBHSA-N
SMILESO[C@@H](CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAFumeric Acid: ≤0.5%
Assay from Supplier's CofAMaleic Acid: ≤0.2%
Assay from Supplier's CofAMalic Acid: ≥98.5% (Titration)
Insoluble material≤0.1%
Residue on ignition≤0.1%
View more
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

Solubility
Soluble in water(363g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Isao Takata; Kozo Yamamoto; Tetsuya Tosa; Ichiro Chibata. Immobilization of Brevibacterium flavum with carrageenan and its application for continuous production of l-malic acid.Enzyme and Microbial Technology.1980, 2, 30-36.
  2. F. A. T. Anet. The Configuration of Deuterio-l-malic acid Produced Enzymatically. Synthesis of Threo-3-deuterio-dl-malic acid. J. Am. Chem. Soc.1960, 82, (4), 994-995.