BCA Protein Assay Kit
Thermo Scientific Chemicals
BCA Protein Assay Kit
製品番号(カタログ番号)数量
J63283.QA1 Kit
製品番号(カタログ番号) J63283.QA
価格(JPY)
-
数量:
1 Kit
一括またはカスタム形式をリクエストする
仕様
概要Clear
使用対象(アプリケーション)Contains two separate solutions for estimation of protein concentration.
製品タイプProtein Assay Kit
数量1 Kit
推奨保存方法Ambient temperatures
さらに表示
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes. Accurately measures protein concentrations from 0.5 to 30 ?g/mL in tube assays and 1 to 20 ?g/mL in 96- or 384-well plate assays. It is used for studying protein: protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
よくあるご質問(FAQ)
What BCA protein assay kits do you recommend for NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C)?
NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C) are compatible with most commercial BCA protein assay kits. We offer a number of BCA protein assay kits, including, but not limited to, the BCA Protein Assay Kit (Cat. No. J63283.QA), Micro BCA Protein Assay Kit (Cat. No. 23235, and Pierce BCA Protein Assay Kit (Cat. No 23225, 23227).
To identify, compare, and choose the best protein assay for your sample type and application, you can use our interactive Protein Assay Selection Guide.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What are the contents of the BCA Protein Assay Kit (Cat. No. J63283.QA)?
The BCA Protein Assay Kit contains 250 mL of Solution A and 5.5 mL of Solution B. The solution components are as follows:
  • Solution A: Bicinchonic acid and tartrate in an alkaline carbonate buffer
  • Solution B: 4% w/v cupric sulfate