RIPA buffer (2X)
Thermo Scientific Chemicals
RIPA buffer (2X)
製品番号(カタログ番号) J63324.AK
または、製品番号J63324-AK
価格(JPY)
-
数量:
250mL
一括またはカスタム形式をリクエストする
仕様
濃度2X
フォーマット液体
数量250mL
推奨保存方法Keep cold
製品タイプCell Lysis Reagent
さらに表示
RIPA is used as a high quality, ready to use mammalian cell and tissue lysis buffer. If protease inhibitors are included after receipt, it can be used to lyse cells and tissue, for radio immunoprecipitation assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
2° to 8°C