BIS-TRIS, 0.2M buffer soln., pH 7.0
BIS-TRIS, 0.2M buffer soln., pH 7.0
BIS-TRIS, 0.2M buffer soln., pH 7.0
Thermo Scientific Chemicals

BIS-TRIS, 0.2M buffer soln., pH 7.0

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
数量:
100 mL
250 mL
製品番号(カタログ番号) J63346.AK
または、製品番号J63346-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
CAS6976-37-0
ChEBICHEBI:41250
化学物質名または材質BIS-TRIS
Colorless
IUPAC名2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only