Ethylenediamine-N,N'-diacetic acid
Ethylenediamine-N,N'-diacetic acid
Ethylenediamine-N,N'-diacetic acid
Thermo Scientific Chemicals

Ethylenediamine-N,N'-diacetic acid

Acts as a chelating agent | CAS: 5657-17-0 | C6H10N2Na2O4 | 220.14 g/mol
製品番号(カタログ番号) J63570.06
または、製品番号J63570-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン1778355
CAS5657-17-0
化学物質名または材質Ethylenediamine-N,N'-diacetic acid
White
EINECS番号227-105-6
さらに表示
Ethylenediamine-N,N'-diacetic acid is acts as a chelating agent, used as a haemostatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine-N,N′-diacetic acid is acts as a chelating agent, used as a haemostatic.

Solubility
Soluble in warm water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Rong Ma.; Ramunas J. Motekaitis.; Arthur E. Martell. Stability of metal ion complexes of N,N'-bis(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid.Inorganica Chimica Acta.1994, (1-2), 151-155.
  2. Ivan Legg, J.; Dean W. Cooke. The Stereochemistry of Some Cobalt(III) Complexes with Ethylenediamine-N,N'-diacetic Acid and Some N-Substituted Analogs.Inorg. Chem.1965, (11), 1576-1584.