1-Aminobenzotriazole, 97%
1-Aminobenzotriazole, 97%
1-Aminobenzotriazole, 97%
Thermo Scientific Chemicals

1-Aminobenzotriazole, 97%

Suicide inhibitor of cytochrome P450 and chloroperoxidase | CAS: 1614-12-6 | C6H6N4 | 134.142 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) J63610.MD
または、製品番号J63610-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン607843
CAS1614-12-6
化学物質名または材質1-Aminobenzotriazole
Brown to White
式量134.14
さらに表示
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

Solubility
Soluble in chloroform, DMSO, ethanol or water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Elizabeth A. Dierks.; Christopher Davis, S.; Paul R. Ortiz de Montellano. Glu-320 and Asp-323 Are Determinants of the CYP4A1 Hydroxylation Regiospecificity and Resistance to Inactivation by 1-Aminobenzotriazole.Biochemistry.1998, (7), 1839-1847.
  2. Miranda, C.L.; Henderson, M.C.; Buhler, D.R. Evaluation of Chemicals as Inhibitors of Trout Cytochrome P450s.Toxicology and Applied Pharmacology.1998, (2), 237-244.