2,9-Dimethyl-1,10-phenanthroline, 98%
2,9-Dimethyl-1,10-phenanthroline, 98%
2,9-Dimethyl-1,10-phenanthroline, 98%
Thermo Scientific Chemicals

2,9-Dimethyl-1,10-phenanthroline, 98%

Reagent for the colorimetric determination of copper | CAS: 484-11-7 | C14H12N2 | 208.264 g/mol
製品番号(カタログ番号) J63698.14
または、製品番号J63698-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS484-11-7
Appearance (Color)White to pale cream to yellow
FormPowder
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)<0.5 %
Melting point158 to 162°C
さらに表示
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ashbrook, L. N.; Elliott, C. M. Dye-Sensitized Solar Cell Studies of a Donor-Appended Bis(2,9-dimethyl-1,10-phenanthroline) Cu(I) Dye Paired with a Cobalt-Based Mediator. J. Phys. Chem. C 2013, 117 (8), 3853-3864.
  2. Xiao, C-L.; Wang, C-Z.; Yuan, L-Y.; Li, B.; He, H.; Wang, S.; Zhao, Y-L.; Chai, Z-F.; Shi, W-Q. Excellent Selectivity for Actinides with a Tetradentate 2,9-Diamide-1,10-Phenanthroline Ligand in Highly Acidic Solution: A Hard-Soft Donor Combined Strategy. Inorg. Chem. 2014, 53 (3), 1712-1720.
  3. Ashbrook, L. N.; Elliott, C. M. Dye-Sensitized Solar Cell Studies of a Donor-Appended Bis(2,9-dimethyl-1,10-phenanthroline) Cu(I) Dye Paired with a Cobalt-Based Mediator. J. Phys. Chem. C. 2013, 117 (8), 3853-3864.