Diethylaminoethyl dextran
Diethylaminoethyl dextran
Diethylaminoethyl dextran
Thermo Scientific Chemicals

Diethylaminoethyl dextran

CAS: 9015-73-0
製品番号(カタログ番号) J63781.14
または、製品番号J63781-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9015-73-0
IUPAC NameDiethylaminoethyl dextran
MDL NumberMFCD00081561
FormPowder
Loss on Drying<10.0%
Total Nitrogen3.0 to 3.8%
pH(aq soln.): 5.0 to 7.0
Viscosity0.4 to 0.6
さらに表示
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. The use of DEAE-Dextran as an efficient adjuvant in vaccine production is well established. For instance, DEAE-Dextran has been used for immunization of guinea pigs and swine with inactivated foot-and-mouth disease and rhesus monkeys with inactivated Venezulean equine encephalomyelitis virus, IVEE. The mechanism of action of DEAE-Dextran is not clear. The effect on the humoral response of rhesus monkeys to IVEE vaccine results in a typical IgM-IgG response. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. James H. McCutchan,; Joseph S. Pagano. Enhancement of the Infectivity of Simian Virus 40 Deoxyribonucleic Acid With Diethylaminoethyl-Dextran2. J Natl Cancer Inst. 1968, 41 (2),351-357..
  2. H Duc-Nguyen. Enhancing effect of diethylaminoethyl-dextran on the focus-forming titer of a murine sarcoma virus (Harvey strain). J Virol. 1968, 2 (6), 643-644.