Sodium phosphate, 0.5M buffer soln., pH 7.0
Thermo Scientific Chemicals

Sodium phosphate, 0.5M buffer soln., pH 7.0

数量:
250 mL
500 mL
製品番号(カタログ番号) J63791.AK
または、製品番号J63791-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
化学物質名または材質Sodium phosphate buffer soln.
MDL番号MFCD00146206
推奨保存方法Ambient temperatures
溶解度情報Fully miscible.
TSCAYes
さらに表示
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

Solubility
Fully miscible.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Masters, C.L.; Simms, G.; Weinman, N.A.; Multhaup, G.; McDonald, B.L.; Beyreuther, K. Amyloid plaque core protein in Alzheimer disease and Down syndrome. Current Issue. 1985, 82, (12), 4245-4249.
  2. Volker M. Vogt. Purification and Further Properties of Single-Strand-Specific Nuclease from Aspergillus oryzae. European Journal of Biochemistry. 1973, 33, (1), 192-200.

よくあるご質問(FAQ)

The specifications for Sodium phosphate, 0.5M buffer soln., pH 7.0 say to store in cool dry conditions. Does this mean I need to store it in the refrigerator?

Sodium phosphate, 0.5M buffer soln., pH 7.0 can be stored at ambient temperature, 15-25° Celsius. It does not need to be stored in a refrigerator.