Hemoglobin, Bovine
Thermo Scientific Chemicals

Hemoglobin, Bovine

For research use | 64.5KDa
製品番号(カタログ番号) J63838.14
または、製品番号J63838-14
価格(JPY)
-
数量:
25g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9008-02-0
MDL NumberMFCD00131282
Appearance (Color)Dark red to brown
FormPowder or crystalline powder
Loss on Drying≤10.0%
pH7.0-9.0
Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide. It is used as a coloring agent for pet foods, a natural source of iron for nutraceuticals, a protein source for non-ruminant animals, and as a raw material for pharmaceutical porphyrin derivative production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only