Sodium citrate, 0.5M buffer soln., pH 6.5
Thermo Scientific Chemicals

Sodium citrate, 0.5M buffer soln., pH 6.5

数量:
250 mL
500 mL
製品番号(カタログ番号) J63888.AK
または、製品番号J63888-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
仕様
化学物質名または材質Sodium citrate buffer soln.
Colorless
MDL番号MFCD00012462
臭気Odorless
推奨保存方法Keep cold
さらに表示
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

Solubility
Soluble in water.

Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.
RUO – Research Use Only

General References:

  1. CJ Quaife.; SD Findley.; JC Erickson.; GJ Froelick. Induction of a new metallothionein isoform (MT-IV) occurs during differentiation of stratified squamous epithelia. Biochemistry. 1994, 33,(23), 7250-7259.
  2. M Cappello.; S Keshav.; C Prince.; DP Jewell.; S Gordon. Detection of mRNAs for macrophage products in inflammatory bowel disease by in situ hybridization. Gut . 1992, 33,(9), 1214-1219.