Thermo Scientific Chemicals

Phorbol 12-myristate 13-acetate, 95%, Thermo Scientific Chemicals

Catalog number: J63916.LB0
5 mg, Each
Thermo Scientific Chemicals

Phorbol 12-myristate 13-acetate, 95%, Thermo Scientific Chemicals

Catalog number: J63916.LB0
5 mg, Each
Quantity
Catalog number: J63916.LB0
also known as J63916-LB0
Price (USD)
Price: 263.00
Online price: 223.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16561-29-8
IUPAC Name
(1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Formula
C36H56O8
InChI Key
PHEDXBVPIONUQT-RGYGYFBISA-N
SMILES
CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥94.0%
Form
Powder, film or foam, solid

Description

PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PMA activates Ca2+- ATPase and potentiates forskolin-induced cAMP formation. It has been shown to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.

Solubility
Very soluble in methanol, soluble in DMSO, chloroform, ethanol or acetone, ether, ethyl acetate, DMF or acetonitrile. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text