Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
Thermo Scientific Chemicals

Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%

Inhibitor of papain and trypsin | CAS: 4238-41-9 | C14H22Cl2N2O3S | 369.30 g/mol
製品番号(カタログ番号) J63959.MC
または、製品番号J63959-MC
価格(JPY)
-
数量:
100 mg
一括またはカスタム形式をリクエストする
仕様
CAS4238-41-9
化学物質名または材質Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride
White
EINECS番号224-199-0
式量369.31
さらに表示
Inhibitor of papain and trypsin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only