2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%
2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%
2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%
Thermo Scientific Chemicals

2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%

A tricyclic norepinephrine uptake inhibitor | CAS: 14028-44-5 | C17H16ClN3O | 313.785 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) J64066.06
または、製品番号J64066-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
CAS14028-44-5
ChEBICHEBI:2675
化学物質名または材質2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine
White
EINECS番号237-867-1
さらに表示
A tricyclic norepinephrine uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only