Laminin (929-933)
Thermo Scientific Chemicals

Laminin (929-933)

594.66 g/mol
製品番号(カタログ番号)数量
J64127.MCR
または、製品番号J64127-M^
1 mg
製品番号(カタログ番号) J64127.MCR
または、製品番号J64127-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110590-64-2
Assay from Supplier's CofAPeptide purity: ≥94.0% (HPLC)
CommentCounter ion: trifluoroacetate
Appearance (Color)White
FormLyophilized powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
-30°C to -10°C