N(epsilon)-Acetyl-L-lysine, 99%
N(epsilon)-Acetyl-L-lysine, 99%
N(epsilon)-Acetyl-L-lysine, 99%
Thermo Scientific Chemicals

N(epsilon)-Acetyl-L-lysine, 99%

CAS: 692-04-6 | C8H16N2O3 | 188.23 g/mol
製品番号(カタログ番号) J64139.03
または、製品番号J64139-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS692-04-6
IUPAC Name(2S)-2-amino-6-acetamidohexanoic acid
Molecular FormulaC8H16N2O3
InChI KeyDTERQYGMUDWYAZ-ZETCQYMHSA-N
SMILESCC(=O)NCCCC[C@H](N)C(O)=O
さらに表示
Appearance (Color)White
Assay (HPLC)> 98.5%
FormPowder
Optical Rotation21.0 to 23.0 (C=2 in 5N HCl)
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml).

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
Sohye Kang; Zhongqi Zhang; Jason Richardson; Bhavana Shah; Shivani Gupta; Chung-Jr Huang; Jinshu Qiu; Nicole Le; Henry Lin; Pavel V.Bondarenko. Metabolic markers associated with high mannose glycan levels of therapeutic recombinant monoclonal antibodies. Journal of Biotechnology. 2015, 203,22-31.