Acycloguanosine, 98%
Acycloguanosine, 98%
Acycloguanosine, 98%
Thermo Scientific Chemicals

Acycloguanosine, 98%

Inhibits cytomegalovirus replication; induces apoptosis | CAS: 59277-89-3 | C8H11N5O3 | 225.21 g/mol
製品番号(カタログ番号) J64144.03
または、製品番号J64144-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS59277-89-3
ChEBICHEBI:2453
化学物質名または材質Acycloguanosine
White
EINECS番号261-685-1
さらに表示
Inhibits cytomegalovirus replication; induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only