alpha-Melanocyte Stimulating Hormone amide
alpha-Melanocyte Stimulating Hormone amide
alpha-Melanocyte Stimulating Hormone amide
Thermo Scientific Chemicals

alpha-Melanocyte Stimulating Hormone amide

An endogenous melanocortin receptor agonist. Anti-inflammatory peptide; antagonizes proinflammatory mediators, including TNF-α, IL-6 and NO and induces anti-inflammatory cytokine IL-10.
製品番号(カタログ番号) J64310.MCR
または、製品番号J64310-M^
価格(JPY)
-
数量:
1 mg
一括またはカスタム形式をリクエストする
仕様
CAS581-05-5
フォーマットSolid
IUPAC名(4S)-4-[(1-{[(1S)-1-{[(1S)-1-{[(1S)-1-[({[(2S)-6-amino-1-[(2S)-2-{[(1S)-1-carbamoyl-2-methylpropyl]carbamoyl}pyrrolidin-1-yl]-1-oxohexan-2-yl]carbamoyl}methyl)carbamoyl]-2-(1H-indol-3-yl)ethyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-2-phenylethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl)carbamoyl]-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-acetamido-3-hydroxypropanamido]-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido]-4-(methylsulfanyl)butanamido]butanoic acid
InChIキーWHNFPRLDDSXQCL-PDEGJNOBSA-N
MDL番号MFCD00133062
さらに表示
alpha-Melanocyte Stimulating Hormone amide is an endogenous melanocortin receptor agonist (Ki values are 0.12, 31, 660 and 5700 nM for MC1, MC3, MC4 and MC5 receptors respectively). Anti-inflammatory peptide; antagonizes proinflammatory mediators, including TNF-α, IL-6 and NO and induces anti-inflammatory cytokine IL-10. Inhibits food intake and induces penile erections following i.c.v. administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
組成および保存条件
Store at -20°C