Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Deoxyribonucleic acid sodium salt, calf thymus, Thermo Scientific Chemicals

Catalog number: J64400.MC
100 mg, Each
Thermo Scientific Chemicals

Deoxyribonucleic acid sodium salt, calf thymus, Thermo Scientific Chemicals

Catalog number: J64400.MC
100 mg, Each
Quantity
Catalog number: J64400.MC
also known as J64400-MC
Price (USD)

Description

Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.

Solubility
Soluble in water.

Notes
Highly polymerized. Hyperchromicity is at least 27%.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text