L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
Solubility
Soluble in water
Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Leonidas Zervas.; Dimitrios M. Theodoropoulos. N-Tritylamino Acids and Peptides. A New Method of Peptide Synthesis1. J. Am. Chem. Soc. 1956, 78, (7), 1359-1363.
- Leonidas. Zervas.; Iphigenia. Photaki. On Cysteine and Cystine Peptides. I. New S-Protecting Groups for Cysteine. J. Am. Chem. Soc. 1962, 84, (20), 3887-3897.