Thermo Scientific Chemicals
Z-Asp-Glu-Val-Asp-7-amino-4-(trifluoromethyl)coumarin
Catalog number: J64569.LB0
5 mg, Each
Thermo Scientific Chemicals
Z-Asp-Glu-Val-Asp-7-amino-4-(trifluoromethyl)coumarin
Catalog number: J64569.LB0
5 mg, Each
Quantity
Catalog number: J64569.LB0
also known as J64569-#0
Price (USD)
Price: 267.00
Online price: 233.65
Your price:
Quantity
-
Description
Z-Asp-Glu-Val-Asp-7-amino-4-(trifluoromethyl)coumarin acts as a substrate for granzyme B-processed ICE-LAP3, caspase-6, caspase-7, caspase-8, and caspase-10. The reaction can be observed by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore.